Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107027.1 | NP_001100497.1 | 3 - 4 | 744 | 153 | |
XM_006246987.2 | XP_006247049.1 | 3 - 4 | 734 | 153 | ||
XM_006246988.2 | XP_006247050.1 | 3 - 4 | 734 | 153 | ||
GenBank mRNA | BC168749.1 | - | 2 - 3 | 213 | 153 | |
FQ225888.1 | - | 2 - 3 | 235 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |