Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246987.2 | XP_006247049.1 | 11 - 12 | 1572 | 83 | |
XM_006246988.2 | XP_006247050.1 | 11 - 12 | 1572 | 83 | ||
GenBank mRNA | BC168749.1 | - | 10 - 11 | 1051 | 83 | |
FQ225888.1 | - | 10 - 11 | 1075 | 83 | ||
FQ230964.1 | - | 6 - 7 | 629 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |