Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 3 - 4 | 382 | 67 | |
XM_006245977.2 | XP_006246039.1 | 3 - 4 | 4200 | 67 | ||
XM_008767578.1 | XP_008765800.1 | 4 - 5 | 3317 | 67 | ||
XM_008767579.1 | XP_008765801.1 | 4 - 5 | 3192 | 67 | ||
XM_008767580.1 | XP_008765802.1 | 3 - 4 | 1302 | 67 | ||
GenBank mRNA | BC099777.1 | - | 3 - 4 | 382 | 7933232 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |