Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107016.1 | NP_001100486.1 | 4 - 5 | 468 | 70 | |
GenBank mRNA | FQ233279.1 | - | 4 - 5 | 340 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |