Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017009.2 | NP_058705.2 | 1 - 2 | 458 | 82 | |
XM_008768261.1 | XP_008766483.1 | 1 - 2 | 524 | 82 | ||
GenBank mRNA | BC088851.1 | - | 1 - 2 | 458 | 7315338 | 82 |
EF094476.1 | - | 1 - 2 | 529 | 82 | ||
L27219.1 | - | 1 - 2 | 462 | 82 | ||
U03700.1 | - | 1 - 2 | 482 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |