Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022601.2 | NP_072123.1 | 4 - 5 | 550 | 67 | |
GenBank mRNA | BC087016.1 | - | 4 - 5 | 550 | 7128621 | 67 |
U91561.1 | - | 4 - 5 | 541 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |