Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007724.1 | NP_001007725.1 | 2 - 3 | 278 | 93 | |
XM_006247532.2 | XP_006247594.1 | 2 - 3 | 527 | 93 | ||
XM_008768293.1 | XP_008766515.1 | 2 - 3 | 325 | 93 | ||
XM_008768294.1 | XP_008766516.1 | 2 - 3 | 643 | 93 | ||
GenBank mRNA | BC079186.1 | - | 2 - 3 | 278 | 7131172 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |