Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013875.1 | NP_001013897.1 | 4 - 5 | 697 | 122 | |
XM_006251204.2 | XP_006251266.1 | 5 - 6 | 794 | 122 | ||
XM_006251205.2 | XP_006251267.1 | 4 - 5 | 584 | 122 | ||
XM_006251206.2 | XP_006251268.1 | 5 - 6 | 627 | 122 | ||
XM_006251207.2 | XP_006251269.1 | 4 - 5 | 585 | 122 | ||
XM_006251208.2 | XP_006251270.1 | 5 - 6 | 734 | 122 | ||
XM_006251209.2 | XP_006251271.1 | 4 - 5 | 777 | 122 | ||
XM_006251210.2 | XP_006251272.1 | 3 - 4 | 573 | 122 | ||
GenBank mRNA | BC081912.1 | - | 4 - 5 | 697 | 7112018 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |