Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031019.1 | NP_112281.1 | 1 - 2 | 155 | 112 | |
GenBank mRNA | X03036.1 | - | 1 - 2 | 153 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |