Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271315.1 | NP_001258244.1 | 199 | 94 | ||
GenBank mRNA | L33840.1 | - | 151 | 94 |
Gene Symbol: LOC103689933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759445.1 | XP_008757667.1 | 2 - 3 | 238 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |