Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008314.1 | NP_001008315.1 | 4 - 5 | 653 | 112 | |
GenBank mRNA | BC083870.1 | - | 4 - 5 | 653 | 7128406 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |