Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199207.1 | NP_954677.1 | 14 - 15 | 1299 | 113 | |
XM_006237147.2 | XP_006237209.1 | 14 - 15 | 1323 | 113 | ||
XM_006237149.2 | XP_006237211.1 | 14 - 15 | 1323 | 113 | ||
GenBank mRNA | AY257251.1 | - | 14 - 15 | 1299 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |