Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199207.1 | NP_954677.1 | 25 - 26 | 2763 | 74 | |
XM_006237147.2 | XP_006237209.1 | 25 - 26 | 2787 | 74 | ||
XM_006237149.2 | XP_006237211.1 | 24 - 25 | 2685 | 74 | ||
GenBank mRNA | AY257251.1 | - | 25 - 26 | 2763 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |