Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025049.1 | NP_001020220.1 | 1 - 2 | 134 | 147 | |
XM_006237379.2 | XP_006237441.1 | 1 - 2 | 674 | 147 | ||
GenBank mRNA | BC079212.1 | - | 1 - 2 | 134 | 7131828 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |