Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107886.1 | NP_001101356.1 | 12 - 13 | 2633 | 74 | |
XM_006237300.2 | XP_006237362.1 | 13 - 14 | 2658 | 74 | ||
XM_006237302.2 | XP_006237364.1 | 11 - 12 | 2556 | 74 | ||
XM_006237303.2 | XP_006237365.1 | 12 - 13 | 2498 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |