Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031515.3 | NP_113703.1 | 2 - 3 | 290 | 130 | |
XM_008763352.1 | XP_008761574.1 | 2 - 3 | 2883 | 130 | ||
XM_008763353.1 | XP_008761575.1 | 3 - 4 | 327 | 130 | ||
XM_008763354.1 | XP_008761576.1 | 3 - 4 | 472 | 130 | ||
XM_008763355.1 | XP_008761577.1 | 2 - 3 | 2883 | 130 | ||
GenBank mRNA | BC126086.1 | - | 3 - 4 | 428 | 8362025 | 130 |
FQ230946.1 | - | 3 - 4 | 471 | 130 | ||
U09793.1 | - | 2 - 3 | 290 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |