Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015005.2 | NP_001015005.2 | 10 - 11 | 1119 | 64 | |
XM_006237518.1 | XP_006237580.1 | 11 - 12 | 1176 | 64 | ||
GenBank mRNA | BC091696.1 | - | 10 - 11 | 1112 | 7301732 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |