Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108280.1 | NP_001101750.1 | 4 - 5 | 496 | 92 | |
XM_006246751.1 | XP_006246813.1 | 3 - 4 | 526 | 92 | ||
XM_006246752.2 | XP_006246814.1 | 3 - 4 | 610 | 92 | ||
XM_006246754.2 | XP_006246816.1 | 3 - 4 | 317 | 92 | ||
XM_008767844.1 | XP_008766066.1 | 2 - 3 | 680 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |