Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108299.1 | NP_001101769.1 | 5 - 6 | 526 | 128 | |
XM_006247369.2 | XP_006247431.1 | 5 - 6 | 599 | 128 | ||
XM_006247370.2 | XP_006247432.1 | 5 - 6 | 547 | 128 | ||
XM_006247371.2 | XP_006247433.1 | 5 - 6 | 507 | 128 | ||
XM_006247372.2 | XP_006247434.1 | 5 - 6 | 354 | 128 | ||
XM_008768085.1 | XP_008766307.1 | 5 - 6 | 596 | 128 | ||
XM_008768086.1 | XP_008766308.1 | 5 - 6 | 600 | 128 | ||
GenBank mRNA | BC168950.1 | - | 5 - 6 | 400 | 9022277 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |