Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107074.1 | NP_001100544.1 | 2 - 3 | 433 | 67 | |
GenBank mRNA | BC167030.1 | - | 2 - 3 | 405 | 7378211 | 67 |
FQ226313.1 | - | 2 - 3 | 435 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |