Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024234.1 | NP_001019405.1 | 1 - 2 | 829 | 143 | |
GenBank mRNA | BC094520.1 | - | 1 - 2 | 829 | 7316275 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |