Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017285.1 | NP_058981.1 | 3 - 4 | 373 | 65 | |
GenBank mRNA | BC084723.1 | - | 2 - 3 | 296 | 7319054 | 65 |
D21800.1 | - | 3 - 4 | 373 | 65 | ||
FQ211229.1 | - | 3 - 4 | 330 | 65 | ||
FQ232780.1 | - | 3 - 4 | 369 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |