Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270544.1 | NP_001257473.1 | 3 - 4 | 517 | 129 | |
NM_001270545.1 | NP_001257474.1 | 3 - 4 | 517 | 129 | ||
NM_017322.2 | NP_059018.1 | 3 - 4 | 517 | 129 | ||
XM_006246317.1 | XP_006246379.1 | 3 - 4 | 513 | 129 | ||
XM_006246318.2 | XP_006246380.1 | 3 - 4 | 465 | 129 | ||
GenBank mRNA | BC061870.1 | - | 3 - 4 | 484 | 5598943 | 129 |
L27111.1 | - | 2 - 3 | 273 | 129 | ||
L27112.1 | - | 2 - 3 | 273 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |