Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004272.1 | NP_001004272.1 | 1 - 2 | 181 | 95 | |
XM_006246747.2 | XP_006246809.1 | 1 - 2 | 237 | 95 | ||
GenBank mRNA | BC079162.1 | - | 1 - 2 | 181 | 7129493 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |