Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246868.2 | XP_006246930.1 | 3 - 4 | 990 | 112 | |
XM_006246869.2 | XP_006246931.1 | 3 - 4 | 990 | 112 | ||
XM_006246870.2 | XP_006246932.1 | 3 - 4 | 990 | 112 | ||
XM_573142.6 | XP_573142.4 | 3 - 4 | 990 | 112 | ||
GenBank mRNA | FQ219135.1 | - | 3 - 4 | 462 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons |