Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109031.1 | NP_001102501.1 | 2 - 3 | 207 | 114 | |
XM_006246393.2 | XP_006246455.1 | 3 - 4 | 313 | 114 | ||
XM_006246394.2 | XP_006246456.1 | 2 - 3 | 428 | 114 | ||
XM_006246395.2 | XP_006246457.1 | 2 - 3 | 426 | 114 | ||
XM_008767703.1 | XP_008765925.1 | 2 - 3 | 537 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |