Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053617.2 | NP_446069.1 | 3 - 4 | 312 | 81 | |
GenBank mRNA | BC091133.1 | - | 3 - 4 | 276 | 81 | |
BC107447.1 | - | 3 - 4 | 305 | 7323668 | 81 | |
DY312674.1 | - | 3 - 4 | 312 | 81 | ||
FQ218610.1 | - | 3 - 4 | 317 | 81 | ||
FQ218635.1 | - | 3 - 4 | 266 | 81 | ||
FQ218700.1 | - | 3 - 4 | 317 | 81 | ||
FQ219505.1 | - | 1 - 2 | 98 | 81 | ||
FQ219645.1 | - | 3 - 4 | 384 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |