Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191922.1 | NP_001178851.1 | 4 - 5 | 1015 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |