Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191652.1 | NP_001178581.1 | 6 - 7 | 888 | 86 | |
XM_006247574.2 | XP_006247636.1 | 6 - 7 | 2231 | 86 | ||
XM_006247576.2 | XP_006247638.1 | 6 - 7 | 784 | 86 | ||
XM_008768349.1 | XP_008766571.1 | 6 - 7 | 846 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |