Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147144.1 | NP_671485.1 | 10 - 11 | 1737 | 120 | |
GenBank mRNA | AF525467.1 | - | 10 - 11 | 1737 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |