Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153301.2 | NP_695213.1 | 3 - 4 | 475 | 122 | |
GenBank mRNA | AF415240.1 | - | 3 - 4 | 483 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |