Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247645.1 | XP_006247707.1 | 2 - 3 | 1773 | 102 | |
XM_006247646.2 | XP_006247708.1 | 2 - 3 | 2605 | 102 | ||
XM_006247647.1 | XP_006247709.1 | 2 - 3 | 1773 | 102 | ||
GenBank mRNA | BC083806.1 | - | 1 - 2 | 157 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |