Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134835.1 | NP_001128307.1 | 1 - 2 | 495 | 61 | |
XM_006247714.2 | XP_006247776.1 | 2 - 3 | 354 | 61 | ||
GenBank mRNA | BC158710.1 | - | 1 - 2 | 495 | 9035021 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |