Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107049.1 | NP_001100519.1 | 1 - 2 | 233 | 65 | |
XM_006247310.2 | XP_006247372.1 | 1 - 2 | 1087 | 65 | ||
GenBank mRNA | BC162060.1 | - | 1 - 2 | 176 | 7115660 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |