Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107277.1 | NP_001100747.1 | 4 - 5 | 797 | 118 | |
XM_006252273.2 | XP_006252335.1 | 3 - 4 | 716 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |