Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012773.2 | NP_036905.2 | 7 - 8 | 5219 | 71 | |
XM_006252344.1 | XP_006252406.1 | 8 - 9 | 5393 | 71 | ||
XM_008770886.1 | XP_008769108.1 | 8 - 9 | 6285 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |