Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013187.1 | NP_001013205.1 | 3 - 4 | 340 | 93 | |
XM_008770873.1 | XP_008769095.1 | 3 - 4 | 340 | 93 | ||
GenBank mRNA | AY724532.1 | - | 3 - 4 | 312 | 93 | |
BC087106.1 | - | 3 - 4 | 340 | 7193157 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |