Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025279.1 | NP_001020450.1 | 1 - 2 | 49 | 93 | |
GenBank mRNA | BC097957.1 | - | 1 - 2 | 49 | 7462731 | 93 |
FQ234004.1 | - | 2 - 3 | 276 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |