Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053600.2 | NP_446052.2 | 3 - 4 | 496 | 94 | |
XM_006239637.2 | XP_006239699.1 | 3 - 4 | 547 | 94 | ||
XM_006239638.2 | XP_006239700.1 | 3 - 4 | 547 | 94 | ||
XM_006239639.2 | XP_006239701.1 | 3 - 4 | 547 | 94 | ||
GenBank mRNA | AB076183.1 | - | 3 - 4 | 496 | 94 | |
AB080086.1 | - | 3 - 4 | 475 | 94 | ||
BC100060.1 | - | 3 - 4 | 477 | 7128839 | 94 | |
CK359698.1 | - | 3 - 4 | 463 | 94 | ||
U64689.1 | - | 3 - 4 | 322 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |