Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199269.1 | NP_954890.1 | 5 - 6 | 897 | 100 | |
XM_008764688.1 | XP_008762910.1 | 4 - 5 | 1686 | 100 | ||
GenBank mRNA | AY371924.1 | - | 5 - 6 | 897 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |