Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106710.1 | NP_001100180.1 | 4 - 5 | 2913 | 90 | |
XM_006239793.2 | XP_006239855.1 | 4 - 5 | 2924 | 90 | ||
XM_008764511.1 | XP_008762733.1 | 4 - 5 | 3656 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |