Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729600.3 | XP_002729646.1 | 2 - 3 | 117 | 64 | |
GenBank mRNA | FQ224256.1 | - | 2 - 3 | 78 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |