Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC168942.1 | - | 2 - 3 | 226 | 8365819 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |