Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108294.1 | NP_001101764.1 | 6 - 7 | 499 | 106 | |
GenBank mRNA | FQ225734.1 | - | 6 - 7 | 503 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |