Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768149.1 | XP_008766371.1 | 15 - 16 | 1510 | 81 | |
GenBank mRNA | FQ210246.1 | - | 15 - 16 | 1478 | 81 | |
FQ217168.1 | - | 15 - 16 | 1474 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |