Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107072.1 | NP_001100542.1 | 2 - 3 | 1151 | 89 | |
XM_008768476.1 | XP_008766698.1 | 2 - 3 | 6308 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |