Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107034.1 | NP_001100504.1 | 4 - 5 | 573 | 92 | |
XM_006247036.2 | XP_006247098.1 | 4 - 5 | 671 | 92 | ||
XM_006247037.2 | XP_006247099.1 | 2 - 3 | 2593 | 92 | ||
XM_008768036.1 | XP_008766258.1 | 4 - 5 | 460 | 92 | ||
XM_008768037.1 | XP_008766259.1 | 1 - 2 | 1272 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |