Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105800.1 | NP_001099270.1 | 11 - 12 | 2346 | 81 | |
XM_006246661.2 | XP_006246723.1 | 10 - 11 | 1861 | 81 | ||
XM_006246662.2 | XP_006246724.1 | 10 - 11 | 1848 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |