Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108709.3 | NP_001102179.2 | 6 - 7 | 552 | 81 | |
NM_001163156.3 | NP_001156628.1 | 6 - 7 | 571 | 81 | ||
XM_006240047.2 | XP_006240109.1 | 5 - 6 | 533 | 81 | ||
XM_006240048.2 | XP_006240110.1 | 6 - 7 | 591 | 81 | ||
XM_006240050.2 | XP_006240112.1 | 3 - 4 | 677 | 81 | ||
XM_008764647.1 | XP_008762869.1 | 5 - 6 | 533 | 81 | ||
XM_008764648.1 | XP_008762870.1 | 6 - 7 | 1006 | 81 | ||
GenBank mRNA | BC168860.1 | - | 6 - 7 | 552 | 7121808 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |