Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139326.2 | NP_647542.1 | 1 - 2 | 99 | 122 | |
GenBank mRNA | BC058443.1 | - | 1 - 2 | 99 | 6919050 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |